(1-Tetradecyl)triphenylphosphonium bromide ≥98.0%

Supplier: TCI America
Warning

Synonyms: Triphenyl(tetradecyl)phosphonium bromide, Tetradecyltriphenylphosphonium bromide

T1506-25G
TCT1506-025GEA 126.4 CAD
TCT1506-025G
(1-Tetradecyl)triphenylphosphonium bromide ≥98.0%
(1-Tetradecyl)triphenylphosphonium bromide

CAS Number: 25791-20-2
MDL Number: MFCD00051857
Molecular Formula: C32H44BrP
Molecular Weight: 539.58
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 94


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Myristyltriphenylphosphonium Bromide, (Tetradecyl)triphenylphosphonium Bromide

Formula: H₃C(CH₂)₁₃P(C₆H₅)₃Br
MW: 539.58 g/mol
Melting Pt: 94 °C
MDL Number: MFCD00051857
CAS Number: 25791-20-2
EINECS: 247-267-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2592b6dc439ca6a46dad494d7a50ce37f896e3af&vid=2592b6dc439ca6a46dad494d7a50ce37f896e3af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR