5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)porphyrin ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)-21H,23H-porphine

T1495-100MG
TCT1495-100MGEA 265.44 CAD
TCT1495-100MG
5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)porphyrin ≥95.0% (by titrimetric analysis)
5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)porphyrin

CAS Number: 127812-08-2
MDL Number: MFCD00191499
Molecular Formula: C52H38N4O12
Molecular Weight: 910.89
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: Deep Green


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)-21H,23H-porphine

Formula: C₅₂H₃₈N₄O₁₂
MW: 910.89 g/mol
MDL Number: MFCD00191499
CAS Number: 127812-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4d70bf783dbaaefc62f4bc6e1b9ef48ea6ca40&vid=5f4d70bf783dbaaefc62f4bc6e1b9ef48ea6ca40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR