Thiazolidine-2,4-dicarboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: Tidiacic, Tetrahydrothiazole-2,4-dicarboxylic acid

T1325-5G
TCT1325-005GEA 203.35 CAD
TCT1325-005G
Thiazolidine-2,4-dicarboxylic acid ≥98.0%
Thiazolidine-2,4-dicarboxylic acid

CAS Number: 30097-06-4
MDL Number: MFCD00145399
Molecular Formula: C5H7NO4S
Molecular Weight: 177.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Tetrahydrothiazole-2,4-dicarboxylic Acid

Formula: C₅H₇NO₄S
MW: 177.1784 g/mol
MDL Number: MFCD00145399
CAS Number: 30097-06-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1eaa29eab9bb7f000fece16f7f6f761e67127290&vid=1eaa29eab9bb7f000fece16f7f6f761e67127290&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR