Tetrabutylammonium dichloroaurate

Supplier: TCI America

T1273-100MG
TCT1273-100MGEA 269.39 CAD
TCT1273-100MG
Tetrabutylammonium dichloroaurate
Tetrabutylammonium dichloroaurate

CAS Number: 50480-99-4
MDL Number: MFCD00059125
Molecular Formula: C16H36AuCl2N
Molecular Weight: 510.34
Form: Crystal
Color: White
Melting point (°C): 83

Formula: C₁₆H₃₆AuCl₂N
Melting Pt: 83 °C
MDL Number: MFCD00059125
CAS Number: 50480-99-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68143f8a84dc5f3337c835673e0c38ed64af8a48&vid=68143f8a84dc5f3337c835673e0c38ed64af8a48&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR