1-Butyn-3-yl-p-toluenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: p-Toluenesulphonic acid 1-Butyn-3-yl Ester, (RS)-1-Methyl-2-propynyl-p-toluenesulphonate, p-Toluenesulfonic acid 1-Butyn-3-yl Ester, But-3-yn-2-yl 4-methylbenzenesulfonate, (RS)-1-Methyl-2-propynyl-p-toluenesulfonate, But-3-yn-2-yl 4-methylbenzenesulphonate

T1224-5G
TCT1224-005GEA 262.75 CAD
TCT1224-005G
1-Butyn-3-yl-p-toluenesulfonate ≥98.0%
1-Butyn-3-yl-p-toluenesulfonate

CAS Number: 53487-52-8
MDL Number: MFCD00060095
Molecular Formula: C11H12O3S
Molecular Weight: 224.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 52


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: p-Toluenesulfonic Acid 1-Butyn-3-yl Ester

Formula: C₁₁H₁₂O₃S
MW: 224.27 g/mol
Melting Pt: 52 °C
MDL Number: MFCD00060095
CAS Number: 53487-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00901384c5406adcc9132c4ccaaefc113bc03d4f&vid=00901384c5406adcc9132c4ccaaefc113bc03d4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR