2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1070-500G T1070-25G T1070-100G
TCT1070-500GEA 519.82 CAD
TCT1070-500G TCT1070-025G TCT1070-100G
2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98.0% (by HPLC, titration analysis)
2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]

CAS Number: 41484-35-9
MDL Number: MFCD00059346
Molecular Formula: C38H58O6S
Molecular Weight: 642.94
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 78

Formula: C₃₈H₅₈O₆S
Melting Pt: 78 °C
MDL Number: MFCD00059346
CAS Number: 41484-35-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e13991dcc4e2f62304c052cdc08c03341f487cdb&vid=e13991dcc4e2f62304c052cdc08c03341f487cdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR