Triphenylphosphine sulfide ≥99.0%

Supplier: TCI America
Warning

T1066-25G T1066-500G T1066-100G
TCT1066-025GEA 97.68 CAD
TCT1066-025G TCT1066-500G TCT1066-100G
Triphenylphosphine sulfide ≥99.0%
Triphenylphosphine sulphide

CAS Number: 3878-45-3
MDL Number: MFCD00004917
Molecular Formula: C18H15PS
Molecular Weight: 294.35
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 163

Formula: C₁₈H₁₅PS
Melting Pt: 163 °C
Density: 1.191 g/cm³ (25 °C)
MDL Number: MFCD00004917
CAS Number: 3878-45-3
EINECS: 223-407-7
UN: 3464
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8bdef6ab14952eb2724efe10ac6eecc71527544&vid=b8bdef6ab14952eb2724efe10ac6eecc71527544&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR