Tris(2-methylphenyl)phosphine ≥97.0%

Supplier: TCI America
Warning

Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine

T1024-5G T1024-25G
TCT1024-005GEA 103.95 CAD
TCT1024-005G TCT1024-025G
Tris(2-methylphenyl)phosphine ≥97.0%
Tris(2-methylphenyl)phosphine

CAS Number: 6163-58-2
MDL Number: MFCD00008514
Molecular Formula: C21H21P
Molecular Weight: 304.37
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 124
Flash Point (°C): 198


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: Tris(2-methylphenyl)phosphine

Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Boiling Pt: 187…192 °C (0.05 mmHg)
Melting Pt: 124 °C
Flash Pt: 198 °C
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e52ae7931133d29028afe6a6ef234ff0c9d086f&vid=1e52ae7931133d29028afe6a6ef234ff0c9d086f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR