3,3',5,5'-Tetramethylbenzidine (TMB) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T1023-1G T1023-5G T1023-25G
TCT1023-001GEA 61.1 CAD
TCT1023-001G TCT1023-005G TCT1023-25G
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98.0% (by HPLC, titration analysis)
3,3',5,5'-Tetramethylbenzidine (TMB)

CAS Number: 54827-17-7 MDL Number: MFCD00007748 Molecular Formula: C16H20N2 Molecular Weight: 240.35 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 170


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials Other Product Names / Synonyms: 4,4'-Bi-2,6-xylidine, 4,4'-Diamino-3,3',5,5'-tetramethylbiphenyl, TMB

Formula: C₁₆H₂₀N₂
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00007748
CAS Number: 54827-17-7
EINECS: 259-364-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e49d9629b74d08c5a0a699b5ac2790dd8dd317b&vid=3e49d9629b74d08c5a0a699b5ac2790dd8dd317b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR