D-(+)-Trehalose, anhydrous ≥98.0%

Supplier: TCI America

T0832-25G T0832-5G
TCT0832-025GEA 212.29 CAD
TCT0832-025G TCT0832-5G
D-(+)-Trehalose, anhydrous ≥98.0%
D(+)-Trehalose

CAS Number: 99-20-7
MDL Number: MFCD00006628
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 205
Specific rotation [a]20/D: 197 deg (C=7, H2O)

Formula: C₁₂H₂₂O₁₁
Melting Pt: 205 °C
Density: 1.53 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006628
CAS Number: 99-20-7
EINECS: 202-739-6
Merck Index: 12,09713
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e8e3fba0b7fc3f525a61d47d1037079b58d962e&vid=1d24dfe14c3ec03aee86e38b3b406d9a1dbe81ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR