Tricosanoic acid 95,0

Supplier: TCI America
Warning

T0402-1G T0402-10G
TCT0402-001GEA 278.21 CAD
TCT0402-001G TCT0402-010G
Tricosanoic acid 95,0
Tricosanoic acid

CAS Number: 2433-96-7
MDL Number: MFCD00002808
Molecular Formula: C23H46O2
Molecular Weight: 354.62
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 80

Formula: C₂₃H₄₆O₂
MW: 354.62 g/mol
Boiling Pt: 399
Melting Pt: 76
Storage Temperature: Refrigerator
MDL Number: MFCD00002808
CAS Number: 2433-96-7
EINECS: 219-419-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e014916fbf1b5af803956db0e084bd95a444815&vid=8fb641c38c990f35e8fa180ff0a1734870606891&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR