1,2,4-Triazole ≥99.0%

Supplier: TCI America
Warning

T0340-500G T0340-25G T0340-100G
TCT0340-500GEA 488.22 CAD
TCT0340-500G TCT0340-025G TCT0340-100G
1,2,4-Triazole ≥99.0%
1,2,4-Triazole

CAS Number: 288-88-0
MDL Number: MFCD00005228
Molecular Formula: C2H3N3
Molecular Weight: 69.07
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 260
Melting point (°C): 121
Flash Point (°C): 170


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Nucleic Acid Synthesis
Other Product Names / Synonyms: Pyrrodiazole

Formula: C₂H₃N₃
Boiling Pt: 260 °C
Melting Pt: 121 °C
Density: 1.394 g/cm³ (25 °C)
Flash Pt: 170 °C
MDL Number: MFCD00005228
CAS Number: 288-88-0
EINECS: 206-022-9
Merck Index: 12,09735
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cad7df7c602297ed7d2b51345ac7b4a41e4224e4&vid=cad7df7c602297ed7d2b51345ac7b4a41e4224e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR