2,4,5-Triamino-6-hydroxypyrimidine sulfate, hydrate ≥90.0% (by titrimetric analysis)

Supplier: TCI America

T0335-25G
TCT0335-025GEA 55.3 CAD
TCT0335-025G
2,4,5-Triamino-6-hydroxypyrimidine sulfate, hydrate ≥90.0% (by titrimetric analysis)
2,4,5-Triamino-6-hydroxypyrimidine sulfate

CAS Number: 35011-47-3 MDL Number: MFCD00012785 Molecular Formula: C4H7N5O Molecular Weight: 239.21 Purity/Analysis Method: 90.0% (T) Form: Crystal Color: Very Pale Yellow


Form: Crystal
Color: Very Pale Yellow

Formula: C₄H₉N₅O₅S
MDL Number: MFCD00012785
CAS Number: 35011-47-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=170f0c0e46e28e3c755d50fe87c4440c005f1334&vid=8d5f7bc880c7a950c397db39d40e31e789a28336&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR