o-Toluonitrile ≥98.0%

Supplier: TCI America

T0306-25G T0306-500G
TCT0306-025GEA 38.9 CAD
TCT0306-025G TCT0306-500G
o-Toluonitrile ≥98.0%
o-Toluonitrile

CAS Number: 529-19-1
MDL Number: MFCD00001792
Molecular Formula: C8H7N
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 205
Flash Point (°C): 82
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2-Cyanotoluene

Formula: C₈H₇N
Boiling Pt: 205 °C
Melting Pt: –13 °C
Density: 1.00 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00001792
CAS Number: 529-19-1
EINECS: 208-451-7
Merck Index: 12,09675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42acc0edd599cf025c970d4d6c15f080876c3715&vid=42acc0edd599cf025c970d4d6c15f080876c3715&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR