L-Threonine 99,0

Supplier: TCI America

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

T0230-25G T0230-500G T0230-100G
TCT0230-025GEA 91.58 CAD
TCT0230-025G TCT0230-500G TCT0230-100G
L-Threonine 99,0
L-Threonine

CAS Number: 72-19-5
MDL Number: MFCD00064270
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -28 deg (C=6, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Thr-OH

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=164366cd8264259d018894e47f0e2e8f2d7190ef&vid=164366cd8264259d018894e47f0e2e8f2d7190ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR