N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 50.0%

Supplier: TCI America

Synonyms: N,N,N,N-Tetrakis(2-hydroxyethyl)ethylenediamine

T0120-25G
TCT0120-025GEA 74.26 CAD
TCT0120-025G
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 50.0%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine

CAS Number: 140-07-8
MDL Number: MFCD00002854
Molecular Formula: C10H24N2O4
Molecular Weight: 236.31
Purity/Analysis Method: >50.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 99


Other Product Names / Synonyms: Ethylenediamine-N,N,N',N'-tetraethanol, N,N,N',N'-Tetra(2-hydroxyethyl)ethylenediamine, 2,2',2'',2'''-Ethylenedinitrilotetraethanol

Formula: C₁₀H₂₄N₂O₄
MW: 236.3086 g/mol
Flash Pt: 99 °C
MDL Number: MFCD00002854
CAS Number: 140-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acdaf827a1fb1ed6923b2753ddafc546a87d09bc&vid=acdaf827a1fb1ed6923b2753ddafc546a87d09bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 50.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR