DL-Sodium lactate 60% in water

Supplier: TCI America

Synonyms: (±)-Lactic acid sodium salt, (±)-2-Hydroxypropionic acid sodium salt, Lactate sodium, DL-Lactic acid sodium salt, (±)-2-Hydroxypropanoic acid sodium salt, Sodium 2-hydroxypropanoate

S0928-500G
TCS0928-500GEA 103.77 CAD
TCS0928-500G
DL-Sodium lactate 60% in water
DL-Sodium lactate

CAS Number: 72-17-3
MDL Number: MFCD00065400
Molecular Formula: C3H6O3
Molecular Weight: 112.06
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: DL-Lactic Acid Sodium Salt, Sodium 2-Hydroxypropionate, 2-Hydroxypropionic Acid Sodium Salt

Formula: H₃CCH(OH)COONa
MW: 112.06
MDL Number: MFCD00065400
CAS Number: 72-17-3
Merck Index: 13,08709
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdd8c24bd31dd0217e0a42218315c851aad03e69&vid=73f53b39ac9e313758c8540336f1c3ddf05a4530&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR