N-Succinimidyl trifluoroacetate ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: TFA-NHS, N-(Trifluoroacetoxy)succinimide, Trifluoroacetic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 2,2,2-trifluoroacetate

S0915-1G
TCS0915-1GEA 186.42 CAD
TCS0915-1G
N-Succinimidyl trifluoroacetate ≥95.0% (by GC)
N-Succinimidyl trifluoroacetate
Formula: C₆H₄F₃NO₄
MW: 211.10 g/mol
Boiling Pt: 100°C
MDL Number: MFCD00595252
CAS Number: 5672-89-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR