Swertiamarin ≥98.0% (by HPLC)

Supplier: TCI America

S0897-25MG
TCS0897-25MGEA 421.54 CAD
TCS0897-25MG
Swertiamarin ≥98.0% (by HPLC)
Swertiamarin

CAS Number: 17388-39-5
MDL Number: MFCD07783984
Molecular Formula: C16H22O10
Molecular Weight: 374.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 114
Specific rotation [a]20/D: -138 deg (C=0.5, EtOH)
Lambda max.: 238 nm (MeOH)Storage Temperature: 0-10°C

Formula: C₁₆H₂₂O₁₀
Melting Pt: 114 °C
MDL Number: MFCD07783984
CAS Number: 17388-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8bdd033901b8fcc5a249e7455886d26a0087129&vid=f8bdd033901b8fcc5a249e7455886d26a0087129&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR