Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

Supplier: TCI America

S0883-100MG S0883-20MG
TCS0883-100MGEA 489.8 CAD
TCS0883-100MG TCS0883-20MG
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

CAS Number: 92921-24-9
MDL Number: MFCD00054978
Molecular Formula: C16H18N2O9S
Molecular Weight: 436.37
Form: Crystal
Color: White
Storage Temperature: <0°C


Other Product Names / Synonyms: Sulfo-SMCC Sodium Salt

Formula: C₁₆H₁₈N₂O₉S
MDL Number: MFCD00054978
CAS Number: 92921-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a181ef05392bb3f15d5139b69aeeeaa005a6224&vid=9a181ef05392bb3f15d5139b69aeeeaa005a6224&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR