Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate hydrate ≥98.0% (by HPLC and gravimetric analysis)

Supplier: TCI America

S0447-1G S0447-200MG
TCS0447-1GEA 346.7 CAD
TCS0447-1G TCS0447-200MG
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate hydrate ≥98.0% (by HPLC and gravimetric analysis)
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate hydrate

CAS Number: 79060-88-1
MDL Number: MFCD00043323
Molecular Formula: C32H12BF24Na
Molecular Weight: 886.21
Purity/Analysis Method: >98.0% (HPLC,W)
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: NaBARF

Formula: C₃₂H₁₂BF₂₄Na.xH₂O
MDL Number: MFCD00043323
CAS Number: 79060-88-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12d2b306330348294c479dee62e3724a3bff63ed&vid=5b399f71c625a560c768f0d49ad57bde7377ce51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,W)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR