Diisopropyl succinate ≥99.0%

Supplier: TCI America

S0269-25ML S0269-500ML
TCS0269-025MLEA 74.26 CAD
TCS0269-025ML TCS0269-500ML
Diisopropyl succinate ≥99.0%
Diisopropyl succinate

CAS Number: 924-88-9
MDL Number: MFCD00059360
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 228
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Succinic Acid Diisopropyl Ester

Formula: C₁₀H₁₈O₄
Boiling Pt: 228 °C
Density: 0.99 (20 °C)
MDL Number: MFCD00059360
CAS Number: 924-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ffe5bce9a8186683b6ca34c274c987287ad3547&vid=3ffe5bce9a8186683b6ca34c274c987287ad3547&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR