Salicylaldehyde ≥98.0%

Supplier: TCI America
Warning

S0004-500G S0004-25G S0004-100G
TCS0004-500GEA 117.95 CAD
TCS0004-500G TCS0004-025G TCS0004-100G
Salicylaldehyde ≥98.0%
Salicylaldehyde

CAS Number: 90-02-8
MDL Number: MFCD00003317
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 196
Melting point (°C): 2
Flash Point (°C): 90
Specific Gravity (20/20): 1.17


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 2-Hydroxybenzaldehyde

Formula: C₇H₆O₂
Boiling Pt: 196 °C
Melting Pt: 2 °C
Density: 1.17 (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00003317
CAS Number: 90-02-8
EINECS: 201-961-0
Merck Index: 13,08405
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b90faa7c4cdbce28bca2309e5e4a1b24e3e3063&vid=df676d763e9b922cc0c995b01e6260c4f0829ab7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR