Resiquimod ≥98% (by HPLC)

Supplier: TCI America

Synonyms: 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848

R0197-100MG R0197-25MG
TCR0197-100MGEA 469.26 CAD
TCR0197-100MG TCR0197-25MG
Resiquimod ≥98% (by HPLC)
Resiquimod
Formula: C₁₇H₂₂N₄O₂
MW: 314.389 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00937759
CAS Number: 144875-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86f79eb394a8b5cecbe5c6ee2f68d2d804da8ee4&vid=86f79eb394a8b5cecbe5c6ee2f68d2d804da8ee4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity(HPLC) min. 98.0 area%
Elemental analysis(Nitrogen) 17.00 to 18.50 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR