(S)-RUCY®-XylBINAP

Supplier: TCI America

R0138-1G R0138-200MG
TCR0138-1GEA 366.88 CAD
TCR0138-1G TCR0138-200MG
(S)-RUCY®-XylBINAP
(S)-RUCY®-XylBINAP

CAS Number: 1312713-89-5
MDL Number: MFCD20922903
Molecular Formula: C71H73ClN2O2P2Ru
Molecular Weight: 1184.84
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: RuCl[(S)-daipena][(S)-xylbinap]

Formula: C₇₁H₇₃ClN₂O₂P₂Ru
MDL Number: MFCD20922903
CAS Number: 1312713-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73486037ce9c83094e94a3fc63958f90fa7c6b7c&vid=36350ef0413cfaa45104777103955810ca7c7566&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR