RuCl2[(S)-dm-segphos®][(S,S)-dpen]

Supplier: TCI America

R0130-1G
TCR0130-1GEA 328.96 CAD
TCR0130-1G TCR0130-200MG
RuCl2[(S)-dm-segphos®][(S,S)-dpen]
RuCl2[(S)-dm-segphos®][(S,S)-dpen]

CAS Number: 944450-46-8
MDL Number: MFCD09753032
Molecular Formula: C60H60Cl2N2O4P2Ru
Molecular Weight: 1107.07
Form: Crystal
Color: Yellow

Formula: C₆₀H₆₀Cl₂N₂O₄P₂Ru
MDL Number: MFCD09753032
CAS Number: 944450-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a39b3dc8fd25857d75eae28b1ae404a07ab98ea&vid=5a39b3dc8fd25857d75eae28b1ae404a07ab98ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR