Quinoxalin-5-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Q0095-1G Q0095-200MG
TCQ0095-1GEA 165.9 CAD
TCQ0095-1G TCQ0095-200MG
Quinoxalin-5-ol ≥98.0% (by GC, titration analysis)
Quinoxalin-5-ol

CAS Number: 17056-99-4
MDL Number: MFCD00972004
Molecular Formula: C8H6N2O
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Boiling point (°C): 184
Melting point (°C): 101
Lambda max.: 250 nm (H2O)


Other Product Names / Synonyms: 5-Hydroxyquinoxaline

Formula: C8H6N2O
Boiling Pt: 184 °C
Melting Pt: 101 °C
MDL Number: MFCD00972004
CAS Number: 17056-99-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dae473ffe791fe0eca6af4bd05912040513ab75b&vid=dae473ffe791fe0eca6af4bd05912040513ab75b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR