4-Quinolinecarbaldehyde ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Q0061-1G Q0061-5G Q0061-25G
TCQ0061-001GEA 61.62 CAD
TCQ0061-001G TCQ0061-005G TCQ0061-25G
4-Quinolinecarbaldehyde ≥97.0% (by GC, titration analysis)
4-Quinolinecarbaldehyde

CAS Number: 4363-93-3
MDL Number: MFCD00006781
Molecular Formula: C10H7NO
Molecular Weight: 157.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 123
Melting point (°C): 50
Flash Point (°C): 110


Other Product Names / Synonyms: Cinchoninaldehyde, 4-Formylquinoline

Formula: C₁₀H₇NO
Boiling Pt: 123 °C
Melting Pt: 50 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006781
CAS Number: 4363-93-3
EINECS: 224-453-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7d02f8753d1facd6675b384b238804302f23d8d&vid=ed6066d5574259b234f74d44d378da9bb87667bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR