1,4-Phenylenediphosphonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

P2303-1G P2303-200MG
TCP2303-1GEA 514.32 CAD
TCP2303-1G TCP2303-200MG
1,4-Phenylenediphosphonic acid ≥98.0% (by HPLC, titration analysis)
1,4-Phenylenediphosphonic acid
Formula: C₆H₈O₆P₂
MDL Number: MFCD00475582
CAS Number: 880-68-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR