2-(1H-Pyrazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P2211-1G P2211-5G
TCP2211-1GEA 129.56 CAD
TCP2211-1G TCP2211-5G
2-(1H-Pyrazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
2-(1H-Pyrazol-3-yl)pyridine

CAS Number: 75415-03-1
MDL Number: MFCD00115151
Molecular Formula: C8H7N3
Molecular Weight: 145.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 124
Lambda max.: 303 nm (CH3CN aq.)


Other Product Names / Synonyms: 3-(2-Pyridyl)pyrazol

Formula: C₈H₇N₃
Melting Pt: 124 °C
MDL Number: MFCD00115151
CAS Number: 75415-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10b8dd9668b42d573afcc240164a28fa5bd0f552&vid=10b8dd9668b42d573afcc240164a28fa5bd0f552&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR