Pararosaniline hydrochloride ≥95.0% (by total nitrogen basis) for biochemical research

Supplier: TCI America

P2052-25G P2052-5G
TCP2052-25GEA 85.32 CAD
TCP2052-25G TCP2052-5G
Pararosaniline hydrochloride ≥95.0% (by total nitrogen basis) for biochemical research
Pararosaniline hydrochloride

[for Biochemical Research] CAS Number: 569-61-9 MDL Number: MFCD00001657 Molecular Formula: C19H17N3 Molecular Weight: 323.82 Purity/Analysis Method: 95.0% (N) Form: Crystal Color: Deep Green Melting point (°C): 270 Lambda max.: 539 nm (EtOH)


Product Categories: Bioscience, Glucuronidase Substrates
Other Product Names / Synonyms: Parafuchsin, Basic Red 9

Formula: C₁₉H₁₈ClN₃
Melting Pt: 270 °C
MDL Number: MFCD00001657
CAS Number: 569-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c11232e296827e3080dd699a67945d2aacb2a8f&vid=be671c8362f1f3ebd21b1b05c1702dc1945c85df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR