Pemirolast potassium ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P1995-1G
TCP1995-1GEA 264.49 CAD
TCP1995-1G
Pemirolast potassium ≥98.0% (by HPLC, titration analysis)
Pemirolast potassium

MDL Number: MFCD01690051
Molecular Formula: C10H8N6O
Molecular Weight: 266.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 317
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 9-Methyl-3-(1H-tetrazol-5-yl)-4H-pyrido[1,2-a]pyrimidin-4-one Potassium Salt

Formula: C₁₀H₇KN₆O
Melting Pt: 317 °C
MDL Number: MFCD01690051
CAS Number: 100299-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=652d8a51a74901dbb86b0544e46d1ef4d9624058&vid=652d8a51a74901dbb86b0544e46d1ef4d9624058&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR