10-Phenyl-9-anthraceneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

P1984-1G
TCP1984-1GEA 157.05 CAD
TCP1984-1G
10-Phenyl-9-anthraceneboronic acid (contains varying amounts of Anhydride)
10-Phenyl-9-anthraceneboronic acid

CAS Number: 334658-75-2
MDL Number: MFCD11111989
Molecular Formula: C20H15BO2
Molecular Weight: 298.15
Form: Crystal
Color: Very Pale Yellow

Formula: C₂₀H₁₅BO₂
MDL Number: MFCD11111989
CAS Number: 334658-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13189d0bc817c7821387d441881be7ffafbf2597&vid=13189d0bc817c7821387d441881be7ffafbf2597&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR