2-Propylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1833-5G
TCP1833-5GEA 121.66 CAD
TCP1833-5G
2-Propylpyrazine ≥98.0% (by GC, titration analysis)
2-Propylpyrazine

CAS Number: 18138-03-9
MDL Number: MFCD00049214
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 174
Specific Gravity (20/20): 0.97

Formula: C₇H₁₀N₂
MW: 122.1700 g/mol
Boiling Pt: 174 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00049214
CAS Number: 18138-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5270b2a21b0835d292d3bf52b52c099aaabb67e&vid=c5270b2a21b0835d292d3bf52b52c099aaabb67e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR