[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel ≥96.0%

Supplier: TCI America

P1738-100MG
TCP1738-100MGEA 52.93 CAD
TCP1738-100MG
[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel ≥96.0%
[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel

CAS Number: 264921-97-3
Molecular Formula: C16H13N3NiO3
Molecular Weight: 353.99
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: Pale Yellow

Formula: C₁₆H₁₃N₃NiO₃
CAS Number: 264921-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a543c0a26519ddaaa3aa33213a0c4014a52a175&vid=2a543c0a26519ddaaa3aa33213a0c4014a52a175&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR