Pyridinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P1637-25G P1637-500G P1637-100G
TCP1637-25GEA 36.34 CAD
TCP1637-25G TCP1637-500G TCP1637-100G
Pyridinium chloride ≥98.0% (by HPLC, titration analysis)
Pyridinium chloride

CAS Number: 628-13-7
MDL Number: MFCD00012802
Molecular Formula: C5H5N
Molecular Weight: 115.56
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Boiling point (°C): 224
Melting point (°C): 147


Other Product Names / Synonyms: Pyridinium Chloride

Formula: C₅H₆ClN
Boiling Pt: 224 °C
Melting Pt: 147 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012802
CAS Number: 628-13-7
EINECS: 211-027-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d3f3a74d07f3a2bd6b286da8a788df6fa680368&vid=2d3f3a74d07f3a2bd6b286da8a788df6fa680368&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR