N-Phenyl-1-anthramine 98,0

Supplier: TCI America

Synonyms: 1-Anilinoanthracene

P1494-500MG
TCP1494-500MGEA 1289.28 CAD
TCP1494-500MG
N-Phenyl-1-anthramine 98,0
N-Phenyl-1-anthramine

CAS Number: 98683-00-2
Molecular Formula: C20H15N
Molecular Weight: 269.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Yellow
Melting point (°C): 162


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1-Anilinoanthracene

Formula: C₂₀H₁₅N
MW: 269.35 g/mol
Melting Pt: 162 °C
CAS Number: 98683-00-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec4f6531486ba69604a3a3eb91fd8bfb18cc3db&vid=8ec4f6531486ba69604a3a3eb91fd8bfb18cc3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR