7-Ethoxy-7-oxoheptanoic acid ≥98.0%

Supplier: TCI America

P1244-5G P1244-25G
TCP1244-005GEA 102.54 CAD
TCP1244-005G TCP1244-025G
7-Ethoxy-7-oxoheptanoic acid ≥98.0%
7-Ethoxy-7-oxoheptanoic acid

CAS Number: 33018-91-6
MDL Number: MFCD00801202
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 162
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Ethyl Hydrogen Pimelate, Pimelic Acid Monoethyl Ester

Formula: C₉H₁₆O₄
Boiling Pt: 162 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00014383
CAS Number: 33018-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=234b2d1eed27ecb77a57344d819ab7b61a1e0ffe&vid=234b2d1eed27ecb77a57344d819ab7b61a1e0ffe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR