Diisobutyl perylenedicarboxylate (mixture of regioisomers) ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Perylenedicarboxylic acid Diisobutyl ester

P1197-5G
TCP1197-5GEA 464.52 CAD
TCP1197-5G
Diisobutyl perylenedicarboxylate (mixture of regioisomers) ≥98.0% (by HPLC)
Diisobutyl perylenedicarboxylate
Formula: C₃₀H₂₈O₄
MW: 452.55 g/mol
MDL Number: MFCD00191681
CAS Number: 2744-50-5
EINECS: 220-379-8

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR