3-Phenyl-2-propyn-1-ol ≥97.0%

Supplier: TCI America

P1147-5G P1147-25G
TCP1147-005GEA 137.14 CAD
TCP1147-005G TCP1147-25G
3-Phenyl-2-propyn-1-ol ≥97.0%
3-Phenyl-2-propyn-1-ol

CAS Number: 1504-58-1
MDL Number: MFCD00040914
Molecular Formula: C9H8O
Molecular Weight: 132.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Flash Point (°C): 110
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 1-Hydroxymethyl-2-phenylacetylene, 3-Phenylpropargyl Alcohol, 1-Phenyl-1-propyn-3-ol

Formula: C₉H₈O
Boiling Pt: 145 °C
Density: 1.06 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00040914
CAS Number: 1504-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eba158888a3120d35fa0062b60e932716724a4c6&vid=eba158888a3120d35fa0062b60e932716724a4c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR