4-Pentylbiphenyl ≥96.0%

Supplier: TCI America

P1133-5ML P1133-25ML
TCP1133-5MLEA 55.3 CAD
TCP1133-5ML TCP1133-025ML
4-Pentylbiphenyl ≥96.0%
4-Pentylbiphenyl

CAS Number: 7116-96-3
MDL Number: MFCD00009503
Molecular Formula: C17H20
Molecular Weight: 224.35
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 167
Flash Point (°C): 110
Specific Gravity (20/20): 0.97

Formula: C₁₇H₂₀
Boiling Pt: 167 °C
Melting Pt: 9.5…10.5 °C
Density: 0.97 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009503
CAS Number: 7116-96-3
EINECS: 230-421-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dd510e0b738e7044f73ca5e115eab9df720f889&vid=2dd510e0b738e7044f73ca5e115eab9df720f889&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR