4-Propenylveratrole ≥98.0%

Supplier: TCI America

P1103-500ML P1103-25ML
TCP1103-500MLEA 252.8 CAD
TCP1103-500ML TCP1103-025ML
4-Propenylveratrole ≥98.0%
4-Propenylveratrole

CAS Number: 93-16-3
MDL Number: MFCD00009282
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 264
Flash Point (°C): 100
Specific Gravity (20/20): 1.05


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 1,2-Dimethoxy-4-(1-propenyl)benzene, O-Methyl Isoeugenol, 4-(1-Propenyl)pyrocatechol Dimethyl Ether

Formula: C₁₁H₁₄O₂
Boiling Pt: 264 °C
Density: 1.05 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00009282
CAS Number: 93-16-3
EINECS: 202-224-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b7ed93b7d83f99a0868de7c5eaa0299e8867c3c&vid=0b7ed93b7d83f99a0868de7c5eaa0299e8867c3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR