(S)-(+)-Prolinol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

P1087-25G P1087-5G
TCP1087-25GEA 333.38 CAD
TCP1087-25G TCP1087-005G
(S)-(+)-Prolinol ≥97.0% (by GC, titration analysis)
(S)-(+)-Prolinol

CAS Number: 23356-96-9
MDL Number: MFCD00005255
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 66
Flash Point (°C): 86
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: 30.5 deg (C=1, Toluene)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-2-(Hydroxymethyl)pyrrolidine, (S)-(+)-2-Pyrrolidinemethanol

Formula: C₅H₁₁NO
Boiling Pt: 66 °C
Density: 1.04 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00005255
CAS Number: 23356-96-9
EINECS: 245-605-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b66aa29f6402b50cae69c3561298f8dbe15a9ca&vid=0b66aa29f6402b50cae69c3561298f8dbe15a9ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR