DL-3-Pyrrolidinol ≥97.0% (by titrimetric analysis)

Supplier: TCI America

P1083-5G P1083-25G
TCP1083-005GEA 157.05 CAD
TCP1083-005G TCP1083-025G
DL-3-Pyrrolidinol ≥97.0% (by titrimetric analysis)
DL-3-Pyrrolidinol

CAS Number: 40499-83-0
MDL Number: MFCD00005256
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 179
Flash Point (°C): 110
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: DL-3-Hydroxypyrrolidine

Formula: C₄H₉NO
Boiling Pt: 179 °C
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005256
CAS Number: 40499-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e09d13701ce307b32c70658663c3c951790be77&vid=5e09d13701ce307b32c70658663c3c951790be77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR