Piperonylonitrile ≥98.0%

Supplier: TCI America

P1074-10G P1074-25G
TCP1074-010GEA 74.42 CAD
TCP1074-010G TCP1074-025G
Piperonylonitrile ≥98.0%
Piperonylonitrile

CAS Number: 4421-09-4
MDL Number: MFCD00005820
Molecular Formula: C8H5NO2
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 94


Other Product Names / Synonyms: 1,3-Benzodioxole-5-carbonitrile, 3,4-Methylenedioxybenzonitrile

Formula: C₈H₅NO₂
Melting Pt: 94 °C
MDL Number: MFCD00005820
CAS Number: 4421-09-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b1ea13ea4cde908e50e7e1d5d619bede2a20291&vid=5b1ea13ea4cde908e50e7e1d5d619bede2a20291&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR