3-Phenoxybenzyl alcohol

Supplier: TCI America
Warning

P0979-25G P0979-500G
TCP0979-025GEA 105.86 CAD
TCP0979-025G TCP0979-500G
3-Phenoxybenzyl alcohol
3-Phenoxybenzyl alcohol

CAS Number: 13826-35-2
MDL Number: MFCD00004636
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 300
Flash Point (°C): 113
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 3-(Hydroxymethyl)diphenyl Ether

Formula: C₁₃H₁₂O₂
Boiling Pt: 300 °C
Density: 1.15 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00004636
CAS Number: 13826-35-2
EINECS: 237-525-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8a947a8a54bb02894cf026c35722674ba1992ae&vid=e8a947a8a54bb02894cf026c35722674ba1992ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR