1-Phenyl-2-butanone ≥95.0%

Supplier: TCI America

P0971-25ML P0971-5ML
TCP0971-25MLEA 583.02 CAD
TCP0971-25ML 76220-204
1-Phenyl-2-butanone ≥95.0%
1-Phenyl-2-butanone

CAS Number: 1007-32-5
MDL Number: MFCD00009315
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 226
Flash Point (°C): 90
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Benzyl Ethyl Ketone

Formula: C₆H₅C(O)CH₂CH₂CH₃
Boiling Pt: 226 °C
Melting Pt: 5 °C
Density: 0.99 (20 °C)
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009315
CAS Number: 1007-32-5
EINECS: 213-752-1

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR