1-(Pentafluorophenyl)ethanol

Supplier: TCI America

Synonyms: (1-Hydroxyethyl)pentafluorobenzene

P0925-5G
TCP0925-005GEA 76.03 CAD
TCP0925-005G
1-(Pentafluorophenyl)ethanol
1-(Pentafluorophenyl)ethanol

CAS Number: 830-50-2
MDL Number: MFCD00004510
Molecular Formula: C8H5F5O
Molecular Weight: 212.12
Form: Crystal
Color: Very Pale Yellow
Flash Point (°C): 87


Other Product Names / Synonyms: (1-Hydroxyethyl)pentafluorobenzene

Formula: C₈H₅F₅O
MW: 212.12 g/mol
Flash Pt: 87 °C
MDL Number: MFCD00004510
CAS Number: 830-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd09bcead5862f4ec9eac0592cec2413c6d5f06b&vid=fd09bcead5862f4ec9eac0592cec2413c6d5f06b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR