PIPES monosodium salt ≥99.0% (by titrimetric analysis)

Supplier: TCI America

P0875-25G
TCP0875-025GEA 157.05 CAD
TCP0875-025G
PIPES monosodium salt ≥99.0% (by titrimetric analysis)
PIPES monosodium salt

[Good's buffer component for biological research]
CAS Number: 10010-67-0
MDL Number: MFCD00065472
Molecular Formula: C8H18N2O6S2
Molecular Weight: 324.34
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White


Product Categories: Bioscience, Buffer-related Reagents
Other Product Names / Synonyms: PIPES 1Na

Formula: C₈H₁₇N₂NaO₆S₂
Storage Temperature: Ambient
MDL Number: MFCD00065472
CAS Number: 10010-67-0
EINECS: 233-005-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ab9fa39eee3f01694dc454bc3552edb2b4d92ea&vid=68b54802c9d91919b0b576be7b90e2c1911c0e77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR