(±)-2-Phenyl-1-propanol ≥98.0%

Supplier: TCI America

Synonyms: 2-Phenylpropan-1-ol

P0813-25ML P0813-500ML
TCP0813-025MLEA 67.94 CAD
TCP0813-025ML TCP0813-500ML
(±)-2-Phenyl-1-propanol ≥98.0%
(±)-2-Phenyl-1-propanol

CAS Number: 1123-85-9
MDL Number: MFCD00004736
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 223
Flash Point (°C): 94
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Hydratropic Alcohol

Formula: C₆H₅CH(CH₃)CH₂OH
MW: 136.19 g/mol
Boiling Pt: 223 °C
Melting Pt: –37 °C
Density: 1.00 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00004736
CAS Number: 1123-85-9
EINECS: 214-379-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ed8a69c92257353cb64c06069f202e9139344b5&vid=0ed8a69c92257353cb64c06069f202e9139344b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR